missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) fluoride, Puratronic, 99.99% (metals basis), Thermo Scientific Chemicals

239.19 € - 827.12 €

Chemical Identifiers

CAS 7782-64-1
Molecular Formula F2Mn
Molecular Weight (g/mol) 92.935
MDL Number MFCD00016222
InChI Key CTNMMTCXUUFYAP-UHFFFAOYSA-L
Synonym manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis
PubChem CID 24528
IUPAC Name difluoromanganese
SMILES F[Mn]F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15407395
View Documents
Thermo Scientific Alfa Aesar
047109.14
25 g
239.19 €
25g
Please to purchase this item. Need a web account? Register with us today!
15417395
View Documents
Thermo Scientific Alfa Aesar
047109.22
100 g
827.12 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Manganese(II) fluoride is used in the production of special kinds of glass and lasers. It reacts with antimony pentafluoride in hydrofluoric acid to prepare a complex, Mn(SbF6)2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-64-1
92.935
CTNMMTCXUUFYAP-UHFFFAOYSA-L
24528
F[Mn]F
F2Mn
MFCD00016222
manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis
difluoromanganese

Specifications

856°C
Red
F2Mn
MFCD00016222
14,5729
Soluble in dilute hydrogen fluoride,concentrated hydrochloric acid and nitric acid; Insoluble in water and alcohol.
F[Mn]F
92.935
92.93
Puratronic™
3.98 g/mL
Manganese(II) fluoride
7782-64-1
Crystalline Powder
MnF2
UN3288
manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis
CTNMMTCXUUFYAP-UHFFFAOYSA-L
difluoromanganese
24528
99.99%
Hygroscopic
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-960-0

missing translation for 'rtecsNumber' : OP0875000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.