Learn More
Manganese(II) bromide hydrate, 98%, Thermo Scientific Chemicals
35.50 € - 450.00 €
Chemical Identifiers
CAS | 10031-20-6 |
---|---|
Molecular Formula | Br2H8MnO4 |
Molecular Weight (g/mol) | 286.81 |
MDL Number | MFCD00149791 |
InChI Key | HHDPJRSXPOGIOP-UHFFFAOYSA-L |
Synonym | manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide |
IUPAC Name | manganese(2+) tetrahydrate dibromide |
SMILES | O.O.O.O.[Mn++].[Br-].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11383758
|
Thermo Scientific Alfa Aesar
040245.14 |
25 g |
35.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11373758
|
Thermo Scientific Alfa Aesar
040245.22 |
100 g |
115.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363758
|
Thermo Scientific Alfa Aesar
040245.36 |
500 g |
450.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis. It can be used in place of palladium in the stille reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10031-20-6 | |
286.81 | |
HHDPJRSXPOGIOP-UHFFFAOYSA-L | |
manganese(2+) tetrahydrate dibromide |
Br2H8MnO4 | |
MFCD00149791 | |
manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide | |
O.O.O.O.[Mn++].[Br-].[Br-] |
Specifications
64.3°C (decomposition) | |
Pink | |
Br2H8MnO4 | |
14,5725 | |
Soluble in water, alcohol | |
O.O.O.O.[Mn++].[Br-].[Br-] | |
286.81 | |
98% | |
Hygroscopic |
10031-20-6 | |
Crystalline Aggregates | |
MFCD00149791 | |
manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide | |
HHDPJRSXPOGIOP-UHFFFAOYSA-L | |
manganese(2+) tetrahydrate dibromide | |
214.76 (Anhydrous) | |
Odorless | |
Manganese(II) bromide hydrate |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
missing translation for 'einecsNumber' : 236-591-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only