missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

26.33 € - 130.65 €

Chemical Identifiers

CAS 6156-78-1
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
Synonym manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
PubChem CID 93021
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10700911
SDS View Documents
Thermo Scientific Acros
205660250
25 g Plastic bottle
26.33 €
25g
Please to purchase this item. Need a web account? Register with us today!
10082740
SDS View Documents
Thermo Scientific Acros
205665000
500 g Plastic bottle
37.39 €
500g
Please to purchase this item. Need a web account? Register with us today!
10723475
SDS View Documents
Thermo Scientific Acros
205660025
2.5 kg Plastic bottle
130.65 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con

GHS Signal Word: Warning

missing translation for 'rtecsNumber' : AI5775000

Product Identifier
  • Manganese(II) acetate tetrahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Manganese(II) acetate tetrahydrate, 99+%, for analysis, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.