missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese aluminide, 99.5% (metals basis), Thermo Scientific™
89.70 € - 390.00 €
Chemical Identifiers
CAS | 12253-13-3 |
---|---|
MDL Number | MFCD00213943 |
Synonym | aluminum, compd. with manganese 3:1, aluminum; manganese, trialumane manganese, aluminum, compound with manganese 3:1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
12253-13-3 | |
MFCD00213943 | |
aluminum, compd. with manganese 3:1, aluminum; manganese, trialumane manganese, aluminum, compound with manganese 3:1 |
Specifications
12253-13-3 | |
MFCD00213943 | |
Insoluble in water. | |
99.5% | |
Manganese aluminide |
Lump | |
aluminum, compd. with manganese 3:1, aluminum; manganese, trialumane manganese, aluminum, compound with manganese 3:1 | |
135.88 | |
(metals basis) |
Safety and Handling
missing translation for 'einecsNumber' : 235-492-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures