Learn More
Manganese, 99+%, powder, -40 mesh, Thermo Scientific Chemicals
62.81 € - 172.86 €
Chemical Identifiers
CAS | 7439-96-5 |
---|---|
Molecular Formula | Mn |
Molecular Weight (g/mol) | 54.94 |
MDL Number | MFCD00011111 |
InChI Key | PWHULOQIROXLJO-UHFFFAOYSA-N |
Synonym | colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds |
PubChem CID | 23930 |
ChEBI | CHEBI:35154 |
IUPAC Name | manganese |
SMILES | [Mn] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10522101
|
Thermo Scientific Acros
317442500 |
250 g | Glass bottle |
62.81 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10551554
|
Thermo Scientific Acros
317440010 |
1 kg | Glass bottle |
172.86 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-96-5 | |
54.94 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
23930 | |
manganese |
Mn | |
MFCD00011111 | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
CHEBI:35154 | |
[Mn] |
Specifications
Black-Brown to Brown-Gray | |
99+% | |
Mn | |
15, 5788 | |
PWHULOQIROXLJO-UHFFFAOYSA-N | |
manganese | |
23930 | |
54.94 | |
Powder |
7439-96-5 | |
Mn | |
MFCD00011111 | |
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds | |
[Mn] | |
54.94 | |
CHEBI:35154 | |
≥99.0% (Mn) | |
Manganese, 99+% |
Safety and Handling
GHS H Statement:
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-105-1
missing translation for 'rtecsNumber' : OO9275000
missing translation for 'tsca' : TSCA
- Manganese, powder, -40 mesh
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : Manganese
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.