missing translation for 'onlineSavingsMsg'
Learn More
Maleate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™ Product Code.: 12972145

Maleate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
BIOPHARMA

Product Code. 12972145
500 mL, Glass Bottle
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 12972145

Brand: Fisher Chemical A 026 500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

Molecular Formula C2H2(COO)22-

Specifications

Packaging Glass Bottle
Quantity 500 mL
Physical Form Liquid
Concentration 1 g/L
Type Single-component Standard Solution
Solvent or Matrix H2O
Chemical Name or Material Maleate
compliance-icons
Product Identifier
  • Single-component Standard Solution for Ion chromatography. Maleate - C2H2(COO)22- 1000mg/l in water (A 026)
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Maleate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.