missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Standard for ICP/MS, 2% HNO3, SPEX CertiPrep™

Product Code. 15245946 Shop All SPEX Certiprep Products
125 mL
Click to view available options
Quantity:
125 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Mg1μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7439-95-4, 7697-37-2, 7732-18-5
Packaging Triple-leached LDPE bottle
Quantity 125 mL
CAS Max % 2.0, 0.0, 98.0
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Density 1.01006 g/cm3 at 20°C
Molecular Formula Mg
MDL Number MFCD00085308
Physical Form Liquid
InChI Key JLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES [Mg]
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
IUPAC Name magnesium
Molecular Weight (g/mol) 24.30
PubChem CID 5462224
ChEBI CHEBI:25107
Solvent or Matrix 2% HNO3
Grade Claritas PPT™
Concentration or Composition (by Analyte or Components) 1mg/L
Chemical Name or Material Magnesium Standard, ICP/MS
Show More Show Less

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Magnesium Standard for ICP/MS, 2% HNO3, SPEX CertiPrep™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.