missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, purum p.a., Honeywell™

Product Code. 15646070
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a., heavy,≥98.0% (based on calcined substance, KT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Quantity 1 kg
Linear Formula MgO
UN Number NONH for all modes of transport
Formula Weight 40.3g/mol
Percent Purity ≥98% (based on calcined substance, KT)
Grade purum p.a.
Chemical Name or Material Magnesium oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.