Learn More
Magnesium oxide, nanopowder, 99+% (metals basis), Thermo Scientific Chemicals
S.A. >7.3m2/g
61.20 € - 686.42 €
Chemical Identifiers
CAS | 1309-48-4 |
---|---|
Molecular Formula | MgO |
Molecular Weight (g/mol) | 40.30 |
MDL Number | MFCD00011109 |
InChI Key | CPLXHLVBOLITMK-UHFFFAOYSA-N |
Synonym | magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag |
PubChem CID | 14792 |
IUPAC Name | oxomagnesium |
SMILES | O=[Mg] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11419970
|
Thermo Scientific Alfa Aesar
044733.14 |
25 g |
61.20 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429970
|
Thermo Scientific Alfa Aesar
044733.22 |
100 g |
185.93 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439970
|
Thermo Scientific Alfa Aesar
044733.36 |
500 g |
686.42 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, as a protective coating in plasma displays, as an oxide barrier in spin-tunneling devices, as an optical material, as a component of tubular construction in heating elements and also in libraries for preserving books. Magnesium oxide microspheres are used in the enrichment method of polycyclic aromatic hydrocarbons.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1309-48-4 | |
40.30 | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
14792 | |
O=[Mg] |
MgO | |
MFCD00011109 | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
oxomagnesium |
Specifications
3600°C | |
1309-48-4 | |
Powder | |
MgO | |
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
CPLXHLVBOLITMK-UHFFFAOYSA-N | |
oxomagnesium | |
14792 | |
1.736 | |
3.58 g/cm3 at 20°C |
2852°C | |
White | |
>99% (Metal basis) | |
MFCD00011109 | |
Soluble in hydrochloric acid and dilute acids. Insoluble in water and ethanol. | |
O=[Mg] | |
40.30 | |
40.3 | |
White powder | |
Magnesium oxide |
Safety and Handling
missing translation for 'einecsNumber' : 215-171-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.