missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride Hexahydrate, ≥99.0%, Reagent Grade, Honeywell™
Reagent Grade,≥99.0%
52.51 € - 249.80 €
Chemical Identifiers
CAS | 7791-18-6 |
---|---|
Molecular Formula | Cl2H12MgO6 |
Molecular Weight (g/mol) | 203.30 |
MDL Number | MFCD00149781 |
InChI Key | DHRRIBDTHFBPNG-UHFFFAOYSA-L |
IUPAC Name | magnesium(2+) hexahydrate dichloride |
SMILES | O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15607920
|
Honeywell Research Chemicals
M0250-500G |
500 g | Poly bottle | ||||||
15697910
|
Honeywell Research Chemicals
M0250-1KG |
1 kg | Poly bottle | ||||||
15617920
|
Honeywell Research Chemicals
M0250-5KG |
5 kg | Poly bottle | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7791-18-6 | |
203.30 | |
DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
Cl2H12MgO6 | |
MFCD00149781 | |
magnesium(2+) hexahydrate dichloride |
Specifications
117°C | |
Colorless | |
Crystalline | |
MgCl2 · 6H2O | |
NONH for all modes of transport | |
O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
203.30 | |
≥99.0% | |
1.57g/cu.cm at 20°C |
7791-18-6 | |
5.0 to 6.5 (at 20°C) | |
Cl2H12MgO6 | |
MFCD00149781 | |
DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
magnesium(2+) hexahydrate dichloride | |
203.3g/mol | |
Odorless | |
Magnesium chloride hexahydrate |
Safety and Handling
missing translation for 'rtecsNumber' : 232-094-6