missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride, 6-Hydrate, Crystal, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15539660
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 7791-18-6
Physical Form Crystals
Quantity 500 g
Molecular Formula Cl2H12MgO6
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]
IUPAC Name magnesium(2+) hexahydrate dichloride
Molecular Weight (g/mol) 203.30
Formula Weight 203.3
Grade Multi-Compendial/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.57kg
Chemical Name or Material Magnesium Chloride, 6-Hydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'