missing translation for 'onlineSavingsMsg'
Learn More

m-Tolylacetylene, 97%, Thermo Scientific Chemicals

40.50 € - 105.00 €

Chemical Identifiers

CAS 766-82-5
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD01318174
InChI Key RENYIDZOAFFNHC-UHFFFAOYSA-N
Synonym 3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene
PubChem CID 136600
IUPAC Name 1-ethynyl-3-methylbenzene
SMILES CC1=CC=CC(=C1)C#C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10259293
View Documents
Thermo Scientific Acros
392660010
1 g Glass bottle
40.50 €
1g
Please to purchase this item. Need a web account? Register with us today!
10360444
View Documents
Thermo Scientific Acros
392660050
5 g Glass bottle
105.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

766-82-5
116.16
RENYIDZOAFFNHC-UHFFFAOYSA-N
136600
CC1=CC=CC(=C1)C#C
C9H8
MFCD01318174
3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene
1-ethynyl-3-methylbenzene

Specifications

766-82-5
100.0
0.9000g/mL
25°C
96% min. (GC)
CH3C6H4C∽CH
MFCD01318174
3-ethynyltoluene, 3-methylphenylacetylene, m-tolylacetylene, 1-ethynyl-3-methyl-benzene, 3'-methylphenyl acetylene, benzene,1-ethynyl-3-methyl, benzene, 1-ethynyl-3-methyl, 3'-methylphenylacetylene, 3-methyl phenylacetylene, 3-tolylacetylene
CC1=CC=CC(=C1)C#C
116.16
116.16
Liquid
96.0
Yellow to Brown
170°C to 175°C
Authentic
C9H8
1.5380 to 1.54
0.9
RENYIDZOAFFNHC-UHFFFAOYSA-N
1-ethynyl-3-methylbenzene
136600
97%
m-Tolylacetylene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

Product Identifier
  • m-Tolylacetylene

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.