missing translation for 'onlineSavingsMsg'
Learn More

m-Nitroacetophenone, 98%, Thermo Scientific Chemicals

25.13 € - 85.43 €

Chemical Identifiers

CAS 121-89-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007259
InChI Key ARKIFHPFTHVKDT-UHFFFAOYSA-N
Synonym 3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
PubChem CID 8494
IUPAC Name 1-(3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10336630
View Documents
Thermo Scientific Acros
128331000
100 g Plastic bottle
25.13 €
100g
Please to purchase this item. Need a web account? Register with us today!
10503681
View Documents
Thermo Scientific Acros
128335000
500 g Plastic bottle
85.43 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-89-1
165.15
ARKIFHPFTHVKDT-UHFFFAOYSA-N
8494
CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]
C8H7NO3
MFCD00007259
3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
1-(3-nitrophenyl)ethanone

Specifications

121-89-1
100.0
201.0°C to 203.0°C
97.5% min. (GC)
O2NC6H4COCH3
07, 288
Solubility in water: insoluble. Other solubilities: soluble in alcohol
CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]
165.15
165.15
Crystalline Powder
97.5
76.0°C to 79.0°C
Authentic
C8H7NO3
MFCD00007259
3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
ARKIFHPFTHVKDT-UHFFFAOYSA-N
1-(3-nitrophenyl)ethanone
8494
98%
m-Nitroacetophenone, 98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 204-504-3

missing translation for 'rtecsNumber' : AM9625000

missing translation for 'tsca' : TSCA

Product Identifier
  • m-Nitroacetophenone

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.