missing translation for 'onlineSavingsMsg'
Learn More

Lutetium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

148.74 € - 579.89 €

Chemical Identifiers

CAS 10099-66-8
MDL Number MFCD00011099
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11343467
SDS View Documents
Thermo Scientific Alfa Aesar
018685.03
1 g
148.74 €
1g
Please to purchase this item. Need a web account? Register with us today!
11333467
SDS View Documents
Thermo Scientific Alfa Aesar
018685.06
5 g
579.89 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lutetium(III) chloride, anhydrous is used as a sputtering target in the preparation of bubble memory device. It is widely used in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10099-66-8
MFCD00011099

Specifications

905°C (sublimation)
MFCD00011099
Soluble in water.
99.9%
3.98 g/mL
Lutetium(III) chloride, Anhydrous
10099-66-8
14,5615
281.33
Hygroscopic
(REO)
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

missing translation for 'einecsNumber' : 233-240-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lutetium(III) chloride, anhydrous, 99.9% (REO), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.