missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles
Buerkle™ Lowering Cable for dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials.
Brand: Buerkle™ 5309-0450
566.09 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Good chemical resistance
- For dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials
- Tensile load max. up to 10kg
Specifications
Lowering Cable | |
Loop | |
4.5 mm |
Copper | |
Dipping Bottles and Immersion Cylinders, Manually Operated Drum Reels | |
50 m |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction