missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Phosphate
Accurate test kits for routine water analysis
Brand: Lovibond™ 147250
Includes:
- Aqua Comparator Housing
- Aqua Comparator Disc
- Phosphate HR Tablets (100 pack)
- 2 x 13.5 mm Sample Cells
- Tablet Crusher
- Instructions
- Carrying Case
Description
Accurate test kits for routine water analysis
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
±5% | |
Aqua comparator™ housing, aqua comparator™ disc, phosphate HR tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case | |
0 to 80mg/L |
Routine water analysis | |
100 | |
Phosphate Test Kit |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- COMPARATOR LOVIBOND PHOSPHATE HR
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
Hazard Statement
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : sodium bisulphate, ammonium chloride, boric acid, ammonium meta-vanadate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction