missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Long Handled PharmaScoop™

Sampling Systems™ Long Handled PharmaScoop™ is ideal for scooping powder when you want to keep your hands clear.

Brand:  Sampling Systems™ A396-1000

Product Code. 13482839

  • 165.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Extra long handle
  • Crevice free construction
  • 316L Stainless Steel
  • Rounded corners for easy cleaning
  • GMP correct
Specifications

Specifications

Stainless Steel (316L)
1000 mL
Polished
600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.