Learn More
BINDER™ Data Logger Software
Binder™ Data Logger Software is configuration and evaluation software suitable for all BINDER™ Data Logger Kits.
Brand:  BINDER™ 8012-0821
Product Code. 15671916
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • LOG ANALYZE software kit, configuration and evaluation software for all BINDER Data Logger Kits (incl. USB data cable)
Specifications
Specifications
Data Logger Software - LOG ANALYZE software kit, configuration and evaluation software for all BINDER Data Logger Kits (incl. USB data cable)
All BINDER Data Logger Kits
Software Kit
USB Data Cable