missing translation for 'onlineSavingsMsg'
Learn More

Lobeline sulfate, 99%, Thermo Scientific Chemicals

41.50 € - 737.00 €

Chemical Identifiers

CAS 134-64-5
Molecular Formula C44H56N2O8S
Molecular Weight (g/mol) 773.00
MDL Number MFCD00135592
InChI Key GRZMOSSVIPFGFF-GNJLJDPWSA-N
Synonym lobeline sulfate, bantron, lobelin sulfate, lobeline sulphate, unii-4cj480v2hp, lobeline, sulfate 2:1 salt, ethanone, 2-6-2-hydroxy-2-phenylethyl-1-methyl-2-piperidinyl-1-phenyl-, 2theta-2alpha,6alpha s-, sulfate 2:1 salt, bis--lobeline ; sulfuric acid, smokeless tn, 2-6-2-hydroxy-2-phenylethyl-1-methylpiperidin-2-yl-1-phenylethanone sulfate
PubChem CID 120636
IUPAC Name 2-[(2R,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethanone;sulfuric acid
SMILES OS(O)(=O)=O.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11413567
View Documents
Thermo Scientific Alfa Aesar
A17326.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11423567
View Documents
Thermo Scientific Alfa Aesar
A17326.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11433567
View Documents
Thermo Scientific Alfa Aesar
A17326.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

134-64-5
773.00
GRZMOSSVIPFGFF-GNJLJDPWSA-N
120636
OS(O)(=O)=O.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1
C44H56N2O8S
MFCD00135592
lobeline sulfate, bantron, lobelin sulfate, lobeline sulphate, unii-4cj480v2hp, lobeline, sulfate 2:1 salt, ethanone, 2-6-2-hydroxy-2-phenylethyl-1-methyl-2-piperidinyl-1-phenyl-, 2theta-2alpha,6alpha s-, sulfate 2:1 salt, bis--lobeline ; sulfuric acid, smokeless tn, 2-6-2-hydroxy-2-phenylethyl-1-methylpiperidin-2-yl-1-phenylethanone sulfate
2-[(2R,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethanone;sulfuric acid

Specifications

134-64-5
MFCD00135592
lobeline sulfate, bantron, lobelin sulfate, lobeline sulphate, unii-4cj480v2hp, lobeline, sulfate 2:1 salt, ethanone, 2-6-2-hydroxy-2-phenylethyl-1-methyl-2-piperidinyl-1-phenyl-, 2theta-2alpha,6alpha s-, sulfate 2:1 salt, bis--lobeline ; sulfuric acid, smokeless tn, 2-6-2-hydroxy-2-phenylethyl-1-methylpiperidin-2-yl-1-phenylethanone sulfate
OS(O)(=O)=O.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1.CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1
773.00
773.01
Lobeline sulfate
C44H56N2O8S
UN1544
GRZMOSSVIPFGFF-GNJLJDPWSA-N
2-[(2R,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethanone;sulfuric acid
120636
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.

missing translation for 'rtecsNumber' : OJ8490170

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.