missing translation for 'onlineSavingsMsg'
Learn More

Lithium Nitrate, Honeywell

Reagent Grade

32.96 € - 246.90 €

Chemical Identifiers

CAS 7790-69-4
Molecular Formula LiNO3
Molecular Weight (g/mol) 68.94
MDL Number MFCD00011094
InChI Key IIPYXGDZVMZOAP-UHFFFAOYSA-N
Synonym lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
PubChem CID 10129889
ChEBI CHEBI:63315
IUPAC Name lithium;nitrate
SMILES [Li+].[O-][N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15692260
View Documents
Honeywell Research Chemicals
227986-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15602270
View Documents
Honeywell Research Chemicals
227986-1KG
1 kg
246.90 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

7790-69-4
68.94
IIPYXGDZVMZOAP-UHFFFAOYSA-N
10129889
lithium;nitrate
LiNO3
MFCD00011094
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
CHEBI:63315
[Li+].[O-][N+]([O-])=O

Specifications

7790-69-4
LiNO3
2722
Soluble in water
[Li+].[O-][N+]([O-])=O
68.94
CHEBI:63315
ReagentPlus™
LiNO3
MFCD00011094
lithium nitrate, nitric acid, lithium salt, lithium nitrate, anhydrous, lithium 1+ ion nitronate, ksc492i7j, lithium nitrate 3-hydrate, lithium 1+ ion nitrate, lithium nitrate, anhydrous 25g, lithium nitrate, ionization buffer solution, specpure
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium;nitrate
10129889
68.95g/mol
Lithium Nitrate
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P301 + P312 + P330-P305 + P351 + P338-P370 + P378

H272-H302-H319

missing translation for 'einecsNumber' : 232-218-9

missing translation for 'rtecsNumber' : QU9330000

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.