missing translation for 'onlineSavingsMsg'
Learn More

Lithium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10743744
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chloride
PubChem CID 433294
ChEBI CHEBI:48607
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1382.0°C
Melting Point 605.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 kg
Assay Percent Range 99% min. (Argentometry)
Linear Formula LiCl
Fieser 01,609; 02,246; 04,298; 05,677; 12,277; 16,194
Merck Index 15, 5584
Solubility Information Solubility in water: 832g/L (20°C). Other solubilities: soluble in alcohols,ether,pyridine,,nitrobenzene and acetone
IUPAC Name lithium;chloride
Formula Weight 42.38
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 1.0% max. (1 g, 105°C, 2 hrs)
Packaging Plastic bottle
Specific Gravity 2.06
Density 2.0600g/mL
Chemical Name or Material Lithium chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Lithium chloride, anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Lithium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'