missing translation for 'onlineSavingsMsg'
Learn More

Lithium Carbonate, Honeywell Fluka™

Product Code. 15661230
Click to view available options
Quantity:
100 g
1 kg
6 x 1 kg
100 kg

Purum,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-13-2
Molecular Formula CLi2O3
Molecular Weight (g/mol) 73.89
MDL Number MFCD00011084
InChI Key XGZVUEUWXADBQD-UHFFFAOYSA-L
Synonym lithium carbonate, dilithium carbonate, lithonate, lithobid, lithane, eskalith, lithotabs, carbonic acid lithium salt, liskonum, lithizine
PubChem CID 11125
ChEBI CHEBI:6504
IUPAC Name dilithium;carbonate
SMILES [Li+].[Li+].[O-]C([O-])=O

Specifications

Quantity 100 kg
Linear Formula Li2CO3
UN Number NONH for all modes of transport
Beilstein 3999191
Formula Weight 73.89g/mol
Percent Purity ≥99%
Grade purum
Chemical Name or Material Lithium Carbonate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'