missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetate dihydrate, Reagent Grade, Honeywell

Product Code. 15657910
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6108-17-4
Molecular Formula C2H7LiO4
Molecular Weight (g/mol) 102.01
MDL Number MFCD00066949
InChI Key IAQLJCYTGRMXMA-UHFFFAOYSA-M
Synonym Acetic acid lithium salt
IUPAC Name lithium(1+) acetate dihydrate
SMILES [Li+].O.O.CC([O-])=O

Specifications

CAS Max % ≤100.00
Physical Form Solid
Quantity 1 kg
Beilstein 3564320
Grade Reagent
Packaging Plastic Bottle
Density 1.3 g/cm3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.