missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Liquid Master

Available in a range of lengths and diameters to give a range of sample volumes. Sampling Systems™ Liquid Master is ideal for taking samples from low and medium viscosity liquids.

Brand:  Sampling Systems™ 5035S-1500

Product Code. 15859331

  • 670.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Product Contact Material Available in: 316L Stainless Steel, Food Grade PTFE/FEP, Polypropylene
  • Easily stripped down for cleaning
  • Single Point and Cross Sectional Sampling
Specifications

Specifications

Liquid Sampler
420 mL
25 mm
Stainless Steel (316L)
Sampling Liquids
1500 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.