missing translation for 'onlineSavingsMsg'
Learn More

Bollé Safety B-Clean Lens Cleaning Products/Kits

Product Code. 12659110
Click to view available options
Product Type:
Anti-Fog Spray
Cleaning Station
Lens Cleaner Spray
Multi-function Dry Cleaning Tissues
Tissue Dispenser
Volume (Metric):
50 mL
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

B-Clean Bollé Safety range of cleaning solutions guarantees optimum cleaning. Alcohol and silicone free, its water-based formula guarantees a perfect cleaning and protection for your spectacles and goggles.

  • Alcohol and silicone free
  • Water-based formula guarantees a perfect cleaning and protection for your spectacles and goggles
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleaning Station
Description Lens Cleaning Accessories
For Use With (Application) For All Types of Ocular
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.