missing translation for 'onlineSavingsMsg'
Learn More

Lead Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™

Product Code. 11430309
Click to view available options
Quantity:
50 g
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pb1000μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7439-92-1, 64742-65-0
Packaging Triple-leached LDPE bottle
CAS Min % 0.5, 99.5
Quantity 50 g
CAS Max % 0.5, 99.5
Certificate of Analysis With Certificate of Analysis.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Molecular Formula Pb
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
SMILES [Pb]
IUPAC Name lead
Molecular Weight (g/mol) 207.20
PubChem CID 5352425
ChEBI CHEBI:27889
Grade Oil/Plasma
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
Base Oil 75 1000μg/g ORG-PB8-2Z
Solvent or Matrix Base Oil 75
Chemical Name or Material Lead Oil Standard
Show More Show Less
compliance-icons
Product Identifier
  • 1000 ug/g Lead in Base Oil 75
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1A
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Lead from Lead Oxide

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'