missing translation for 'onlineSavingsMsg'
Learn More

Lead plate, 6.35mm (0.25in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11307447
Click to view available options
Quantity:
15 x 15 cm
30 x 30 cm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-92-1
Molecular Formula Pb
Molecular Weight (g/mol) 207.20
MDL Number MFCD00134050
InChI Key WABPQHHGFIMREM-UHFFFAOYSA-N
Synonym plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant
PubChem CID 5352425
ChEBI CHEBI:27889
IUPAC Name lead
SMILES [Pb]

Specifications

Color Gray
Quantity 30 x 30 cm
Percent Purity 99.9%
Odor Odorless
Weight ≈1.6kg/15x15cm
Form Plate
Assay (metals basis)
Chemical Name or Material Lead plate, 6.35mm (0.25 in.) thick
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead plate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.