Learn More
Lead(II) zirconium oxide, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
85.40 € - 969.00 €
Chemical Identifiers
CAS | 12060-01-4 |
---|---|
Molecular Formula | O3PbZr |
Molecular Weight (g/mol) | 346.421 |
MDL Number | MFCD00016275 |
InChI Key | RSCSNKLWUCUPDE-UHFFFAOYSA-N |
Synonym | lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate |
PubChem CID | 16212542 |
IUPAC Name | dioxido(oxo)zirconium;lead(2+) |
SMILES | [O-][Zr](=O)[O-].[Pb+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11385047
|
Thermo Scientific Alfa Aesar
040754.09 |
10 g |
85.40 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11375047
|
Thermo Scientific Alfa Aesar
040754.18 |
50 g | |||||||
15446345
|
Thermo Scientific Alfa Aesar
040754.30 |
250 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12060-01-4 | |
346.421 | |
RSCSNKLWUCUPDE-UHFFFAOYSA-N | |
16212542 | |
[O-][Zr](=O)[O-].[Pb+2] |
O3PbZr | |
MFCD00016275 | |
lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate | |
dioxido(oxo)zirconium;lead(2+) |
Specifications
12060-01-4 | |
O3PbZr | |
UN3077 | |
RSCSNKLWUCUPDE-UHFFFAOYSA-N | |
dioxido(oxo)zirconium;lead(2+) | |
16212542 | |
99.997% | |
Odorless | |
(metals basis) |
Powder | |
MFCD00016275 | |
lead zirconate, o3zr.pb, dioxido oxo zirconium;lead 2+, $l^ 2-lead 2+ ion oxozirconiumbis olate | |
[O-][Zr](=O)[O-].[Pb+2] | |
346.421 | |
346.42 | |
Puratronic™ | |
7.0 g/mL | |
Lead(II) zirconium oxide |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 235-039-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only