Learn More
Lead(II) sulfate, Reagent Grade, Thermo Scientific Chemicals
For Use in Pigments, Galvanic Batteries, Lithography
45.12 € - 584.00 €
Chemical Identifiers
CAS | 7446-14-2 |
---|---|
Molecular Formula | O4PbS |
Molecular Weight (g/mol) | 303.256 |
MDL Number | MFCD00011166 |
InChI Key | KEQXNNJHMWSZHK-UHFFFAOYSA-L |
Synonym | lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite |
PubChem CID | 24008 |
IUPAC Name | lead(2+);sulfate |
SMILES | [O-]S(=O)(=O)[O-].[Pb+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11490051
|
Thermo Scientific Alfa Aesar
044784.22 |
100 g |
45.12 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400061
|
Thermo Scientific Alfa Aesar
044784.36 |
500 g |
165.83 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480051
|
Thermo Scientific Alfa Aesar
044784.A4 |
2.5 kg | N/A | ||||||
Description
Lead(II) sulfate is used in pigments, galvanic batteries, lithography, weighting fabrics. It is also used as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7446-14-2 | |
303.256 | |
KEQXNNJHMWSZHK-UHFFFAOYSA-L | |
24008 | |
[O-]S(=O)(=O)[O-].[Pb+2] |
O4PbS | |
MFCD00011166 | |
lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite | |
lead(2+);sulfate |
Specifications
1170°C | |
White | |
O4PbS | |
lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite | |
KEQXNNJHMWSZHK-UHFFFAOYSA-L | |
lead(2+);sulfate | |
24008 | |
Reagent | |
White powder/lump | |
Lead(II) sulfate |
7446-14-2 | |
Lump/Powder | |
MFCD00011166 | |
Partially soluble in water,dilute HCl and HNO3. Soluble in sodium hydroxide,ammonium acetate or tartrate,concentrated hydriodic acid. Insoluble in alcohol | |
[O-]S(=O)(=O)[O-].[Pb+2] | |
303.256 | |
303.25 | |
Odorless | |
6.2 g/cm3 at 20°C |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
H302+H312+H332-H350-H360-H373
missing translation for 'dotInformation' : DOT Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 231-198-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) sulfate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.