Learn More
Lead(II) oxide, low silver, Thermo Scientific Chemicals
31.20 € - 66.90 €
Chemical Identifiers
CAS | 1317-36-8 |
---|---|
Molecular Formula | OPb |
Molecular Weight (g/mol) | 223.20 |
MDL Number | MFCD00011164 |
InChI Key | YEXPOXQUZXUXJW-UHFFFAOYSA-N |
Synonym | lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow |
PubChem CID | 14827 |
SMILES | O=[Pb] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337658
|
Thermo Scientific Alfa Aesar
033330.36 |
500 g |
31.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11347658
|
Thermo Scientific Alfa Aesar
033330.A3 |
2 kg |
66.90 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lead(II) oxide is used in storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores
Chemical Identifiers
1317-36-8 | |
223.20 | |
YEXPOXQUZXUXJW-UHFFFAOYSA-N | |
14827 |
OPb | |
MFCD00011164 | |
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow | |
O=[Pb] |
Specifications
886°C | |
Powder | |
MFCD00011164 | |
14,5413 | |
Insoluble in water and alcohol; Soluble in acetic acid,dilute HNO3 and alkalies | |
O=[Pb] | |
223.20 | |
223.19 | |
9.53 g/mL |
1317-36-8 | |
OPb | |
UN3077 | |
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow | |
YEXPOXQUZXUXJW-UHFFFAOYSA-N | |
plumbanone | |
14827 | |
Odorless | |
Lead(II) oxide, Low silver |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H372
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 215-267-0
missing translation for 'rtecsNumber' : OG1750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Lead(II) oxide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only