missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Nitrate Solution, Fisher Chemical™
BIOPHARMA

Product Code. 12943554
1000 mL
Click to view available options
Quantity:
1000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Quantity 1000 mL
Concentration or Composition (by Analyte or Components) 0.01mol/L
compliance-icons
Product Identifier
  • Lead nitrate 0.01M Volumetric Solution
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lead(II) Nitrate Solution, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.