missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) Nitrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10335760
Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
Synonym lead dinitrate, lead nitrate, lead ii nitrate, plumbous nitrate, lead 2+ nitrate, lead nitrate pb no3 2, nitrate de plomb french, lead ii nitrate 1:2, nitric acid, lead 2+ salt
PubChem CID 24924
ChEBI CHEBI:37187
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 470°C
Color White
pH 3 to 4
Physical Form Solid
Quantity 500 g
IUPAC Name lead(2+);dinitrate
Formula Weight 331.2g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead(II) nitrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P281-Use personal protective equipment as required.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'