Learn More
Lead(II) fluoride, 99% min, Thermo Scientific Chemicals
27.00 € - 300.00 €
Chemical Identifiers
CAS | 7783-46-2 |
---|---|
Molecular Formula | F2Pb |
Molecular Weight (g/mol) | 245.20 |
MDL Number | MFCD00011162 |
InChI Key | FPHIOHCCQGUGKU-UHFFFAOYSA-L |
Synonym | lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure |
PubChem CID | 24549 |
IUPAC Name | difluorolead |
SMILES | [F-].[F-].[Pb++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379229
|
Thermo Scientific Alfa Aesar
088282.18 |
50 g | |||||||
11369229
|
Thermo Scientific Alfa Aesar
088282.30 |
250 g |
93.70 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389229
|
Thermo Scientific Alfa Aesar
088282.A1 |
1 kg |
300.00 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lead(II) fluoride is a specialty product for proteomics research. It is used in low melting glasses, in glass coatings to reflect infrared rays, in phosphors for television-tube screens and as a catalyst for the manufacture of picoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-46-2 | |
245.20 | |
FPHIOHCCQGUGKU-UHFFFAOYSA-L | |
24549 | |
[F-].[F-].[Pb++] |
F2Pb | |
MFCD00011162 | |
lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure | |
difluorolead |
Specifications
1290°C | |
7783-46-2 | |
F2Pb | |
UN2291 | |
lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure | |
[F-].[F-].[Pb++] | |
245.20 | |
245.2 | |
Odorless | |
Lead (II) fluoride |
855°C | |
Powder | |
MFCD00011162 | |
14,5408 | |
FPHIOHCCQGUGKU-UHFFFAOYSA-L | |
difluorolead | |
24549 | |
≥99% | |
8.445g/mL |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
missing translation for 'tsca' : TSCA
- Lead(II) fluoride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only