missing translation for 'onlineSavingsMsg'
Learn More

Lead(Ii) Carbonate Basic, Honeywell Fluka™

Product Code. 15601060
Click to view available options
Quantity:
1 kg
50 kg
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss., white lead,≥77% Pb basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1319-46-6
MDL Number MFCD00078155

Specifications

Quantity 50 kg
Linear Formula (PbCO3)2·Pb(OH)2
UN Number 3077
Formula Weight 775.63g/mol
Percent Purity ≥77% (Pb basis)
Grade puriss.
Chemical Name or Material Lead(Ii) Carbonate Basic
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'