missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lead(II) Bromide, Pure, Fisher Chemical™
23.70 € - 44.40 €
Chemical Identifiers
CAS | 10031-22-8 |
---|---|
Molecular Formula | Br2Pb |
Molecular Weight (g/mol) | 367.00 |
MDL Number | MFCD00011156 |
InChI Key | ZASWJUOMEGBQCQ-UHFFFAOYSA-L |
Synonym | lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x |
PubChem CID | 24831 |
IUPAC Name | λ²-lead(2+) dibromide |
SMILES | [Br-].[Br-].[Pb++] |
Description
Chemical Identifiers
10031-22-8 | |
367.00 | |
ZASWJUOMEGBQCQ-UHFFFAOYSA-L | |
24831 | |
[Br-].[Br-].[Pb++] |
Br2Pb | |
MFCD00011156 | |
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x | |
λ²-lead(2+) dibromide |
Specifications
916°C | |
10031-22-8 | |
Powder/Solid | |
MFCD00011156 | |
ZASWJUOMEGBQCQ-UHFFFAOYSA-L | |
λ²-lead(2+) dibromide | |
24831 | |
Plastic powder jar |
373°C | |
White | |
Br2Pb | |
lead ii bromide, lead bromide, lead dibromide, lead bromide pbbr2, pbbr2, lead 2+ bromide, dibromoplumbane, ccris 4220, leadbromide, acmc-1bq2x | |
[Br-].[Br-].[Pb++] | |
367.00 | |
367.01g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Lead(II) bromide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title