missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Lead Acetate Test Paper

Provides for the quick and easy determination of lead hydrogen sulphide in samples. Macherey Nagel™ Lead Acetate Test Paper is toxic and needs to be declared as hazardous.

Brand:  Macherey-Nagel™ 90744


Product Code. 11386534

  • 15.60 € / 5m

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Provides for the quick and easy determination of lead hydrogen sulphide in samples. Macherey Nagel™ Lead Acetate Test Paper is toxic and needs to be declared as hazardous.

  • Qualitative
  • Sulfide containing solutions also give a positive reaction
  • Non-toxic and safe sulfide test paper is an alternative
Specifications

Specifications

White to Brown/Black
Determination of lead hydrogen sulphide
Qualitative Test Paper
5000 mm
Reel
1 Each
5000 x 7 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lead acetate paper

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360D-May damage the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : lead(II) acetate (trihydrate)

SDS
Documents

Documents

Certificates
Special Offers

Special Offers