Learn More
Trajan Scientific and Medical™ LC PEEKsil™ Tubing
Mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems.
Brand:  Trajan Scientific and Medical™ 0624226
 View more versions of this product
Product Code. 15288733
  • 104.00 € / Pack of 5
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
Specifications
Specifications
Tubing
1/16 in.
1.6 mm
Polymer-sheathed Fused Silica
5/Pk.
0.025 mm
100 mm