missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ BioClean Alpha™ Sterile Latex Gloves

Product Code. 15248088
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides exceptional flexibility and is anatomically shaped for enhanced wearer comfort. Nitritex™ BioClean Alpha™ Sterile Latex Gloves features an extra-thick beaded cuff adds strength and stability on the arm, and reduces the risk of the gloves tearing when donning.

  • Extra thick beaded cuff to reduce tearing when donning
  • Powder-free EasyOn™ proprietary technology allows for easy double-donning
  • Non-particulating EasyTear™ packaging
  • Mid-arm 300mm (12 in.) length provides extra protection
  • Latex gloves offer exceptional flexibility and comfort
  • Anatomically-shaped, hand-specific gloves for a close fit
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) ISO 9001:2015, (EU)2016/425, EN420:2003 +A1:2009 and EN374-1:2016, EN374-5:2016 and EN421:2010, EN455-2:2015, ISO 11137-1:2006, (EC)1935/2004 and (EC) 2023/2006
Cleanroom Class Class 10 (ISO 4)
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Grade EU GMP Grade A
Hand Specific
Material Latex
Powdered Powder Free
Product Type Cleanroom Gloves
Size 6
Sterility Sterile
Glove Cuff Style Beaded/Rolled
Packaging Type Bagged
Quantity One pair per inner PE wallet; one wallet per sealed EasyTear PE pouch; 10 pouches per sealed outer PE bag; 20 outer bags per lined carton (200 pairs)
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'