missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14651-1-AP-20UL
This item is not returnable.
View return policy
Description
Lamina-associated polypeptide 2 (LAP2), isoforms alpha/beta/gamma is a protein that in humans is encoded by the TMPO gene. LAP2 is an inner nuclear membrane (INM) protein. LAP2 plays a role in the regulation of nuclear architecture by binding lamin B1 and chromosomes. This interaction is regulated by phosphorylation during mitosis.Specifications
LAP2 | |
Polyclonal | |
Unconjugated | |
TMPO | |
CMD1T, LAP2, LEMD4, PRO0868, Splenin Thymopentin, thymopoietin, Thymopoietin isoform alpha, TMPO, TP, TP alpha, TP5, TPRP isoform alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21917, 25359, 7112 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P42166, Q61033, Q62733 | |
Tmpo | |
LAP2 Fusion Protein Ag6247 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction