missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanum(III) sulfate hydrate, 99.9% (REO), Thermo Scientific Chemicals
85.30 € - 351.00 €
Chemical Identifiers
CAS | 57804-25-8 |
---|---|
Molecular Formula | La2O12S3 |
Molecular Weight (g/mol) | 565.98 |
MDL Number | MFCD00011070 |
InChI Key | VQEHIYWBGOJJDM-UHFFFAOYSA-H |
IUPAC Name | dilanthanum(3+) trisulfate |
SMILES | [La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11374768
|
Thermo Scientific Alfa Aesar
043106.22 |
100 g |
85.30 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11384768
|
Thermo Scientific Alfa Aesar
043106.36 |
500 g |
351.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as colorant or laboratory reagent. It is also mainly applied in specialty glass, phosphors, water treatment and catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57804-25-8 | |
565.98 | |
VQEHIYWBGOJJDM-UHFFFAOYSA-H | |
[La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
La2O12S3 | |
MFCD00011070 | |
dilanthanum(3+) trisulfate |
Specifications
White heat (decomposition) | |
White | |
99.9% (REO) | |
MFCD00011070 | |
VQEHIYWBGOJJDM-UHFFFAOYSA-H | |
dilanthanum(3+) trisulfate | |
566.00 (anhydrous) | |
Hygroscopic | |
Lanthanum(III) sulfate hydrate |
57804-25-8 | |
Crystalline | |
La2O12S3 | |
Slightly soluble in water. | |
[La+3].[La+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
565.98 | |
Odorless | |
2.821 |
Safety and Handling
missing translation for 'einecsNumber' : 233-239-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only