Learn More
Lanthanum(III) chloride heptahydrate, ACS, Thermo Scientific Chemicals
68.34 € - 142.71 €
Chemical Identifiers
CAS | 10025-84-0 |
---|---|
Molecular Formula | Cl3H14LaO7 |
Molecular Weight (g/mol) | 371.36 |
MDL Number | MFCD00149756 |
InChI Key | FDFPDGIMPRFRJP-UHFFFAOYSA-K |
IUPAC Name | lanthanum(3+) heptahydrate trichloride |
SMILES | O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377647
|
Thermo Scientific Alfa Aesar
012916.18 |
50 g |
68.34 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367647
|
Thermo Scientific Alfa Aesar
012916.30 |
250 g |
142.71 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lanthanum chloride solutions are used in various water treatment applications. Lanthanum chloride is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-84-0 | |
371.36 | |
FDFPDGIMPRFRJP-UHFFFAOYSA-K | |
O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3] |
Cl3H14LaO7 | |
MFCD00149756 | |
lanthanum(3+) heptahydrate trichloride |
Specifications
91°C (decomposition) | |
Crystalline | |
MFCD00149756 | |
Soluble in ethanol and water. | |
O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3] | |
371.36 | |
ACS |
10025-84-0 | |
Cl3H14LaO7 | |
14,5363 | |
FDFPDGIMPRFRJP-UHFFFAOYSA-K | |
lanthanum(3+) heptahydrate trichloride | |
371.37 (245.27 Anhydrous) | |
Lanthanum(III) chloride heptahydrate, For analysis ACS |
Safety and Handling
GHS H Statement
H315-H319-H335
P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c
H290-H317-H318
missing translation for 'einecsNumber' : 233-237-5
missing translation for 'rtecsNumber' : OE4470000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.