Learn More
Lanthanum(III) carbonate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
101.51 € - 391.95 €
Chemical Identifiers
CAS | 54451-24-0 |
---|---|
Molecular Formula | C3La2O9 |
Molecular Weight (g/mol) | 457.84 |
MDL Number | MFCD00011069 |
InChI Key | NZPIUJUFIFZSPW-UHFFFAOYSA-H |
IUPAC Name | dilanthanum(3+) tricarbonate |
SMILES | [La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11302298
|
Thermo Scientific Alfa Aesar
013197.18 |
50 g |
101.51 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312298
|
Thermo Scientific Alfa Aesar
013197.30 |
250 g |
391.95 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lanthanum(III) carbonate hydrate is used as a starting material for the preparation of lanthanum strontium manganite, which is used in solid oxide fuel cell applications. It is also involved in the preparation of high-temperature superconductors. It finds application in medicine as a phosphate binder. Further, it is used in the treatment of hyperphosphatemia. In addition to this, it is used for the tinting of glass, water treatment and as a catalyst for hydrocarbon cracking.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54451-24-0 | |
457.84 | |
NZPIUJUFIFZSPW-UHFFFAOYSA-H | |
[La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
C3La2O9 | |
MFCD00011069 | |
dilanthanum(3+) tricarbonate |
Specifications
54451-24-0 | |
C3La2O9 | |
14,5364 | |
NZPIUJUFIFZSPW-UHFFFAOYSA-H | |
dilanthanum(3+) tricarbonate | |
457.85 (Anhydrous) | |
Reacton™ | |
2.6 g/mL to 2.7 g/mL | |
Lanthanum(III) carbonate hydrate |
Lump/Powder | |
MFCD00011069 | |
Soluble in acids. Insoluble in water. | |
[La+3].[La+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O | |
457.84 | |
99.99% | |
Hygroscopic | |
(REO) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 209-599-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.