missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11322298
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Lanthanum(III) bromide is used as a scintillation material in certain applications. It is used in gamma spectroscopy radioisotope detection. It is used as a source of lanthanum in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 224183-16-8
Molecular Formula Br3La
Molecular Weight (g/mol) 378.62
MDL Number MFCD00798151
InChI Key XKUYOJZZLGFZTC-UHFFFAOYSA-K
Synonym lanthanum iii bromide hydrate, acmc-1chtf, lanthanum bromide hydrate, lanthanum bromide-water 1/3/1
PubChem CID 71317322
SMILES [Br-].[Br-].[Br-].[La+3]

Specifications

Color White
Physical Form Crystalline
Quantity 250 g
Solubility Information Soluble in water.
IUPAC Name lanthanum(3+);tribromide;hydrate
Formula Weight 378.62 (Anhydrous)
Percent Purity 99.99%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Lanthanum(III) bromide hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.