missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum Chloride 10% Solution, for AAS, Fisher Chemical™

Product Code. 10477450
Click to view available options
Quantity:
250 mL
1 L

Chemical Identifiers

MDL Number 149756

Specifications

pH 1
Physical Form Liquid
Quantity 1 L
Assay Percent Range ≥9.5 and ≤10.5% w/v
Packaging Amber glass bottle
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'