missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum boride, 99.5% (REO), Thermo Scientific Chemicals

91.25 € - 1435.14 €

Chemical Identifiers

CAS 12008-21-8
Molecular Formula B6La-2
Molecular Weight (g/mol) 203.765
MDL Number MFCD00151350
InChI Key IBDHMAYMLSYGAK-UHFFFAOYSA-N
Synonym lanthanum boride, lanthanum hexaboride, lanthanum boride reo, lanthanum hexaboride lab6 grade a, lanthanum hexaboride lab6 grade b, lanthanum hexaboride lab6 grade c, lanthanum hexaboride, powder, 10 mum, lanthanum boride, powder,-325 mesh trace metals basis, pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide lanthanum, line position and line shape standard for powder diffraction, nist srm 660c, lanthanum hexaboride powder
PubChem CID 71308229
SMILES B12B3[B-]14B5[B-]23B45.[La]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11373888
View Documents
Thermo Scientific Alfa Aesar
040325.09
10 g
91.25 €
10g
Please to purchase this item. Need a web account? Register with us today!
11393888
View Documents
Thermo Scientific Alfa Aesar
040325.18
50 g
319.59 €
50g
Please to purchase this item. Need a web account? Register with us today!
11383888
View Documents
Thermo Scientific Alfa Aesar
040325.30
250 g
1435.14 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Lanthanum boride is employed in hot cathodes, which enact and includes electron microscopes, microwave tubes, electron lithography, electron beam welding, X-ray tubes, and free electron lasers. It is administered as a size or strain standard in X-ray powder diffraction to calibrate instrumental broadening of diffraction peaks. It exercises as a superconductor. It has a great potential as a thermionic emitter in renewable energy applications. It is widely used in teleran, electronic industry, instrument, medical equipment, metallurgy and environmental protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12008-21-8
203.765
IBDHMAYMLSYGAK-UHFFFAOYSA-N
71308229
B6La-2
MFCD00151350
lanthanum boride, lanthanum hexaboride, lanthanum boride reo, lanthanum hexaboride lab6 grade a, lanthanum hexaboride lab6 grade b, lanthanum hexaboride lab6 grade c, lanthanum hexaboride, powder, 10 mum, lanthanum boride, powder,-325 mesh trace metals basis, pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide lanthanum, line position and line shape standard for powder diffraction, nist srm 660c, lanthanum hexaboride powder
B12B3[B-]14B5[B-]23B45.[La]

Specifications

2210°C
Powder
MFCD00151350
Insoluble in water,HCl.
B12B3[B-]14B5[B-]23B45.[La]
71308229
99.5%
2.61 g/mL
Lanthanum boride
12008-21-8
B6La-2
lanthanum boride, lanthanum hexaboride, lanthanum boride reo, lanthanum hexaboride lab6 grade a, lanthanum hexaboride lab6 grade b, lanthanum hexaboride lab6 grade c, lanthanum hexaboride, powder, 10 mum, lanthanum boride, powder,-325 mesh trace metals basis, pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide lanthanum, line position and line shape standard for powder diffraction, nist srm 660c, lanthanum hexaboride powder
IBDHMAYMLSYGAK-UHFFFAOYSA-N
203.765
203.78
Odorless
(REO)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-531-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.