Learn More
Brady™ Laminated Polyester GHS Symbol Signs
Help ensure safety at work and maximize productivity with these GHS warning labels. Brady™ Laminated Polyester GHS Symbol Signs are available in a variety of sizes, with and without text. Standardized images reduce the risk of misunderstandings.
Brand: Brady™ 811696
Description
- Self-adhesive diamond shape labels
- Pre-printed with GHS hazard symbols
- Laminated polyester, gloss finish black and red on white
Specifications
ISO 7010, RoHS compliant using EU Directive 2002/95/EC | |
Gloss | |
250/Pk. | |
Permanent Acrylic, Pressure-sensitive | |
Pipe markers and safety signs | |
0.024 mm | |
-40°C to 110°C | |
Compressed Gas | |
Red on White |
PIC 1804-50*50-B7541-RLL-C | |
Polyester | |
Continuous roll with 250 symbols (not labels) per roll | |
Laminated Polyester | |
50 mm | |
50 mm | |
Diamond | |
GHS Symbol Sign |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.