Learn More
Thermo Scientific™ Lambda Exonuclease (10 U/μL)
Lambda Exonuclease is a highly processive exodeoxyribonuclease that selectively digests the 5'-phosphorylated strand of double-stranded DNA in 5'-3' direction.
102.00 € - 386.00 €
Specifications
Concentration 10 U/μL
Enzyme Exonuclease
Product Type Exonuclease
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10520081
View Documents Promotion Details
Thermo Scientific™
EN0561
1000 U
102.00 €
1000 units
10344530
View Documents Promotion Details
Thermo Scientific™
EN0562
5000 U
386.00 €
5000 units
Description
Description
Thermo Scientific Lambda Exonuclease is a highly processive 5'≥3' exodeoxyribonuclease. It selectively digests the 5'-phosphorylated strand of double-stranded DNA. The enzyme exhibits low activity on single-stranded DNA and non-phosphorylated DNA, and has no activity at nicks and limited activity at gaps in DNA.
Highlights
  • Active in Thermo Scientific PCR buffers
Specifications
Specifications
10 U/μL
Exonuclease
Exonuclease