missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LACTB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16783-1-AP-20UL
This item is not returnable.
View return policy
Description
Endoribonuclease; cleaves preferentially 3′ to purine-pyrimidine dinucleotide motifs in single-stranded RNA. The cleavage product contains a free 3′ -OH group. Has no activity with double-stranded RNA or DNA. Required for normal mitochondrial function and cell viability.Specifications
LACTB2 | |
Polyclonal | |
Unconjugated | |
LACTB2 | |
Beta lactamase like protein 2, CGI 83, lactamase, beta 2, LACTB2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
212442, 297768, 51110 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q53H82, Q561R9, Q99KR3 | |
LACTB2 | |
LACTB2 Fusion Protein Ag10213 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |