Learn More
Lab Vision™ Tris Buffered Saline and Tween 20 (20X), Epredia™
Take advantage of a wash buffer for immunolabeling techniques with the Utilize Epredia™ Lab Vision™ Tris-Buffered Saline and Tween™ 20 (20X).
Brand: Epredia TA-125-TT
This item is not returnable.
View return policy
Description
- For in vitro diagnostic use
- Tween 20 is added to promote more effective washings, resulting in decreased nonspecific background staining
- Tween 20 aids reagent spreading on slides
Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week
Specifications
TBS with Tween (TBST) | |
Anatomical Pathology | |
Room Temperature |
125 mL | |
20X |
Product Content Correction
Product Title
Lab Vision™ Tris Buffered Saline and Tween 20 (20X), Epredia™ > 125 mL
For in vitro diagnostic use
Spot an opportunity for improvement?Share a Content Correction