missing translation for 'onlineSavingsMsg'
Learn More

Insulin Receptor (β-subunit) Ab-5 Mouse Monoclonal Antibody, Epredia™

Epredia™ Insulin Receptor (β-Subunit) Ab-5, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-635-P

 View more versions of this product

Product Code. 12681847

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The human insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide-linked subunits in a Β-α-α-Β configuration. The Β-subunit (95kDa) possesses a single transmembrane domain with tyrosine kinase acivity, whereas the α-subunit (135kDa) is completely extracellular.

Host Species: Mouse

Clone: CT-1

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rat.

Epitope: aa-GRVLTLPRS- within the C-terminus

Immunogen: A 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor β-subunit

Molecular Weight: 95kDa

Positive Control: IM-9 lymphocyte cells

Cellular Localization: Membrane Ab-5 reacts with autophosphorylated and unoccupied receptors as well as the receptor-insulin complex. Ab-5 does not affect insulin-stimulated autophosphorylation.

Recommended for:

  • Immunoprecipitation
  • Affinity Purification (With peptide elution)
  • ELISA (As capture antibody only)
  • Tyrosine Kinase Assay (Ab-mediated capture on microplates)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Insulin Receptor (beta-Subunit) Ab-5
Monoclonal
200μg/mL
Mouse
1 mL
Steroids Hormones and Growth Factors
Human, Mouse, Rat
IgG2a κ
Immunoprecipitation
CT-1
Unconjugated
A 15-mer synthetic peptide corresponding to aa Tyr-KKNGRILTLPRSNPS from the C-terminal of human insulin receptor Beta-subunit
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers